missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxysuccinimide, 98+%

Product Code. 11414073
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414073 500 g 500g
11494063 25 g 25g
11404073 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414073 Supplier Thermo Scientific Alfa Aesar Supplier No. A10312.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6066-82-6 | C4H5NO3 | 115.088 g/mol

N-Hydroxysuccinimide is used for improved amidations in the carbodiimide method. It is also used to activate a carboxyl group and reacts with amine to form amide. It is involved in the preparation of N-hydroxymaleimide-styrene copolymer. Further, it finds use in analytical chemistry. As an additive, it is used in the carbodiimide method for improved amidations and peptide couplings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Hydroxysuccinimide is used for improved amidations in the carbodiimide method. It is also used to activate a carboxyl group and reacts with amine to form amide. It is involved in the preparation of N-hydroxymaleimide-styrene copolymer. Further, it finds use in analytical chemistry. As an additive, it is used in the carbodiimide method for improved amidations and peptide couplings.

Solubility
Soluble in water, dimethyl formamide, alcohols and ethyl acetate. Insoluble in cold ether.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and strong bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6066-82-6
Molecular Formula C4H5NO3
Molecular Weight (g/mol) 115.088
MDL Number MFCD00005516
InChI Key NQTADLQHYWFPDB-UHFFFAOYSA-N
Synonym n-hydroxysuccinimide, hosu, 1-hydroxysuccinimide, 2,5-pyrrolidinedione, 1-hydroxy, 1-hydroxy-2,5-pyrrolidinedione, succinimide, n-hydroxy, n-hydroxysuccinimde, n-hydroxysuccinimid, unii-mje3791m4t, ccris 2604
PubChem CID 80170
IUPAC Name 1-hydroxypyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)O

Specifications

Melting Point 94°C to 99°C
Assay Percent Range >98%
Quantity 500 g
Beilstein 113913
Sensitivity Hygroscopic
Solubility Information Soluble in water,dimethyl formamide,alcohols and ethyl acetate. Insoluble in cold ether.
Formula Weight 115.09
Percent Purity ≥98%
Chemical Name or Material N-Hydroxysuccinimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.