missing translation for 'onlineSavingsMsg'
Learn More

N'-Hydroxypyridine-4-carboximidamide, 97%, Thermo Scientific™

Product Code. 10638550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10638550 5 g Glass bottle 5g
10566820 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10638550 Supplier Thermo Scientific Acros Supplier No. 432480050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1594-57-6
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00125873
InChI Key ZUUATXHRJFHSGO-UHFFFAOYSA-N
Synonym n'-hydroxypyridine-4-carboximidamide, pyridine-4-amidoxime, 4-pyridylamidoxime, n-hydroxyisonicotinamidine, n-hydroxy-isonicotinamidine, isonicotinamidoxime, isonicotinohydroxamamide, 4-pyridinecarboxamidoxime, n-hydroxyisonicotinimidamide, z-n'-hydroxyisonicotinimidamide
PubChem CID 5355713
IUPAC Name nitroso(1H-pyridin-4-ylidene)methanamine
SMILES N\C(=N/O)C1=CC=NC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 175.0°C to 180.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 137.14
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material N'-Hydroxypyridine-4-carboximidamide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Pyridylamidoxime
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.