missing translation for 'onlineSavingsMsg'
Learn More

N'-Hydroxypyridine-3-carboximidamide, 97%, Thermo Scientific™

Product Code. 10406974
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10406974 5 g 5g
10203332 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10406974 Supplier Thermo Scientific Maybridge Supplier No. S01204DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1594-58-7
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00265955
InChI Key AQBMQGDKWIPBRF-UHFFFAOYSA-N
Synonym 3-pyridylamidoxime, 3-pyridinecarboxamidoxime, z-n'-hydroxynicotinimidamide, n-hydroxynicotinimidamide, z-n'-hydroxypyridine-3-carboximidamide, n-hydroxy-nicotinamidine, 3-pyridinecarboxamidoxine, 3-pyridinecarboxamide oxime, nicotinamidoxime, hydroxyimino-3-pyridylmethylamine
PubChem CID 5372334
SMILES N\C(=N/O)C1=CC=CN=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
IUPAC Name (Z)-N'-hydroxypyridine-3-carboximidamide
Formula Weight 137.14
Percent Purity 97%
Chemical Name or Material N'-hydroxypyridine-3-carboximidamide
compliance-icons
Product Identifier
  • Pyridin-3-carboxamide oxime
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.