missing translation for 'onlineSavingsMsg'
Learn More

N-(Hydroxymethyl)benzamide, 98%

Product Code. 11459000
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
Unit Size:
100g
10g
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459000 100 g 100g
11429000 10 g 10g
11449000 25 g 25g
11439000 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11459000 Supplier Thermo Scientific Alfa Aesar Supplier No. A12250.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6282-02-6 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6282-02-6
MDL Number MFCD00014415

Specifications

Melting Point 94°C to 96°C
Molecular Formula C9H9ClO
Quantity 100 g
Beilstein 2045129
Synonym 4'-chloropropiophenone, 1-4-chlorophenyl propan-1-one, p-chloropropiophenone, 4-chloropropiophenone, 1-propanone, 1-4-chlorophenyl, propiophenone, 4'-chloro, propiophenone, p-chloro, ethyl p-chlorophenyl ketone, usaf ek-5296, unii-26ay8wja1e
InChI Key ADCYRBXQAJXJTD-UHFFFAOYSA-N
SMILES CCC(=O)C1=CC=C(C=C1)Cl
IUPAC Name 1-(4-chlorophenyl)propan-1-one
Molecular Weight (g/mol) 168.62
PubChem CID 22697
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material N- (Hydroxymethyl)benzamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.