missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Hydroxybenzenecarboximidamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | C6H5C(NH2)=NOH |
Quantity | 25 g |
IUPAC Name | (Z)-N'-hydroxybenzenecarboximidamide |
Formula Weight | 136.15 |
Percent Purity | 97% |
Chemical Name or Material | N'-Hydroxybenzenecarboximidamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'