missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy Sertraline, TRC

Product Code. 30400261
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400261 100 mg -
30408087 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400261 Supplier Toronto Research Chemicals Supplier No. TRCH953730100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124345-07-9
Molecular Formula C17 H17 Cl2 N O
Molecular Weight (g/mol) 322.23
Synonym (1S-cis)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-hydroxy-N-methyl-1-naphthalenamine, N-Hydroxysertraline
IUPAC Name [(2R,5S)-5-acetyloxy-3-benzoyloxyoxolan-2-yl]methyl benzoate
SMILES N(C)(O)[C@@H]1C=2C([C@@H](CC1)C3=CC(Cl)=C(Cl)C=C3)=CC=CC2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H17Cl2NO/c1-20(21)17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11/h2-6,8,10,12,17,21H,7,9H2,1H3/t12-,17-/m0/s1
Formula Weight 321.0687
Chemical Name or Material N-Hydroxy Sertraline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.