missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy Riluzole, TRC

Product Code. 30395945
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395945 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395945 Supplier Toronto Research Chemicals Supplier No. TRCH9533301MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Hydroxy Riluzole
CAS 179070-90-7
Molecular Formula C8 H5 F3 N2 O2 S
Quantity 1 mg
Synonym 2-Benzothiazolamine, N-hydroxy-6-(trifluoromethoxy)-, 2(3H)-Benzothiazolone, 6-(trifluoromethoxy)-, oxime (9CI), N-Hydroxy-6-(trifluoromethoxy)-2-benzothiazolamine, 2-Hydroxyamino-6-(trifluoromethoxy)benzothiazole, N-Hydroxyriluzole
InChI Formula InChI=1S/C8H5F3N2O2S/c9-8(10,11)15-4-1-2-5-6(3-4)16-7(12-5)13-14/h1-3,14H,(H,12,13)
SMILES ONc1nc2ccc(OC(F)(F)F)cc2s1
IUPAC Name N-[6-(trifluoromethoxy)-1,3-benzothiazol-2-yl]hydroxylamine
Molecular Weight (g/mol) 250.2
Formula Weight 250.0024
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.