missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy Pomalidomide, TRC

Product Code. 30389677
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389677 50 mg
30389378 100 mg
30404114 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389677 Supplier Toronto Research Chemicals Supplier No. TRCH95322350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497147-11-2
Molecular Formula C13H11N3O5
Molecular Weight (g/mol) 289.24
Synonym 1H-Isoindole-1,3(2H)-dione, 2-(2,6-dioxo-3-piperidinyl)-4-(hydroxyamino)- (9CI, ACI), 2-(2,6-Dioxo-3-piperidinyl)-4-(hydroxyamino)-1H-isoindole-1,3(2H)-dione (ACI), 2-(2,6-dioxo-3-piperidyl)-4-(hydroxyamino)isoindoline-1,3-dione, 2-(2,6-Dioxo-3-piperidinyl)-4-(hydroxyamino)-1H-isoindole-1,3(2H)-dione, 1H-Isoindole-1,3(2H)-dione, 2-(2,6-dioxo-3-piperidinyl)-4-(hydroxyamino)- (9CI, ACI), 2-(2,6-Dioxo-3-piperidinyl)-4-(hydroxyamino)-1H-isoindole-1,3(2H)-dione (ACI), 2-(2,6-dioxo-3-piperidyl)-4-(hydroxyamino)isoindoline-1,3-dione
IUPAC Name 2-(2,6-dioxopiperidin-3-yl)-4-(hydroxyamino)isoindole-1,3-dione
SMILES ONc1cccc2C(=O)N(C3CCC(=O)NC3=O)C(=O)c12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H11N3O5/c17-9-5-4-8(11(18)14-9)16-12(19)6-2-1-3-7(15-21)10(6)13(16)20/h1-3,8,15,21H,4-5H2,(H,14,17,18)
Formula Weight 289.0699
Chemical Name or Material N-Hydroxy Pomalidomide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.