missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy Palbociclib, TRC

Product Code. 30403233
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403233 50 mg 50mg
30390122 10 mg 10mg
30398580 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403233 Supplier Toronto Research Chemicals Supplier No. TRCH93997550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2231085-26-8
Molecular Formula C24H29N7O3
Molecular Weight (g/mol) 463.53
Synonym 6-Acetyl-8-cyclopentyl-2-[[5-(4-hydroxy-1-piperazinyl)-2-pyridinyl]amino]-5-methyl-pyrido[2,3-d]pyrimidin-7(8H)-one
IUPAC Name 6-acetyl-8-cyclopentyl-2-[[5-(4-hydroxypiperazin-1-yl)pyridin-2-yl]amino]-5-methylpyrido[2,3-d]pyrimidin-7-one
SMILES CC(=O)C1=C(C)c2cnc(Nc3ccc(cn3)N4CCN(O)CC4)nc2N(C5CCCC5)C1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H29N7O3/c1-15-19-14-26-24(27-20-8-7-18(13-25-20)29-9-11-30(34)12-10-29)28-22(19)31(17-5-3-4-6-17)23(33)21(15)16(2)32/h7-8,13-14,17,34H,3-6,9-12H2,1-2H3,(H,25,26,27,28)
Formula Weight 463.2332
Chemical Name or Material N-Hydroxy Palbociclib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.