missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy Norfloxacin, TRC

Product Code. 30010390
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010390 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30010390 Supplier Toronto Research Chemicals Supplier No. TRCH9488105MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 109142-49-6
Molecular Formula C16 H18 F N3 O4
Quantity 5 mg
InChI Formula InChI=1S/C16H18FN3O4/c1-2-18-9-11(16(22)23)15(21)10-7-12(17)14(8-13(10)18)19-3-5-20(24)6-4-19/h7-9,24H,2-6H2,1H3,(H,22,23)
SMILES CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCN(O)CC3
IUPAC Name 1-ethyl-6-fluoro-7-(4-hydroxypiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
Molecular Weight (g/mol) 335.33
Formula Weight 335.1281
Chemical Name or Material N-Hydroxy Norfloxacin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.