missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy Mexiletine Oxalate, TRC

Product Code. 30405666
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405666 25 mg
30397131 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30405666 Supplier Toronto Research Chemicals Supplier No. TRCH94802525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57204-78-1
Molecular Formula C13H19NO6
Molecular Weight (g/mol) 285.29
Synonym (+/-)-1-(2,6-Dimethylphenoxy)-N-hydroxy-2-propanamine Ethanedioate
IUPAC Name N-[1-(2,6-dimethylphenoxy)propan-2-yl]hydroxylamine;oxalic acid
SMILES CC(COc1c(C)cccc1C)NO.OC(=O)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H17NO2.C2H2O4/c1-8-5-4-6-9(2)11(8)14-7-10(3)12-13;3-1(4)2(5)6/h4-6,10,12-13H,7H2,1-3H3;(H,3,4)(H,5,6)
Formula Weight 285.1212
Chemical Name or Material N-Hydroxy Mexiletine Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.