missing translation for 'onlineSavingsMsg'
Learn More

N'-Hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzenecarboximidamide, 95%, Thermo Scientific™

Product Code. p-2706277
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10542652 250 mg Amber Glass Bottle 250mg
10009391 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10542652 Supplier Thermo Scientific Maybridge Supplier No. JM00487CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-19-3
Molecular Formula C12H13N3O2S
Molecular Weight (g/mol) 263.315
MDL Number MFCD00830779
InChI Key QHRPKGPTUYZTBY-UHFFFAOYSA-N
Synonym n'-hydroxy-4-2-methyl-1,3-thiazol-4-yl methoxy benzenecarboximidamide, 4-2-methylthiazol-4-ylmethoxy benzamidoxime, n'-hydroxy-4-2-methylthiazol-4-yl methoxy benzimidamide, hydroxyimino 4-2-methyl 1,3-thiazol-4-yl methoxy phenyl methylamine, z-n'-hydroxy-4-2-methyl-1,3-thiazol-4-yl methoxy benzene-1-carboximidamide, n'-hydroxy-4-2-methyl-1,3-thiazol-4-yl methoxy benzene-1-carboximidamide
PubChem CID 9582192
IUPAC Name N'-hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzenecarboximidamide
SMILES CC1=NC(=CS1)COC2=CC=C(C=C2)C(=NO)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 263.32
Percent Purity 95%
Chemical Name or Material N'-Hydroxy-4-[(2-methyl-1, 3-thiazol-4-yl)methoxy]benzenecarboximidamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.