missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy-2-methylpropanimidamide, 97%, Thermo Scientific™

Product Code. 10045112
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10045112 10 g Amber Glass Bottle 10g
10085062 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10045112 Supplier Thermo Scientific Maybridge Supplier No. MO07460EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 35613-84-4
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
MDL Number MFCD03426251,MFCD08705910,MFCD00464225
InChI Key JHRDEHLFNLLCQS-UHFFFAOYSA-N
Synonym z-n'-hydroxyisobutyrimidamide, 1z-n'-hydroxy-2-methylpropanimidamide, n-hydroxyisobutyrimidamide, n-hydroxy-2-methylpropanimidamide, z-n'-hydroxy-2-methylpropimidamide, n-hydroxyisobutyramidine, z-n'-hydroxy-2-methylpropanimidamide, isobutyramidoxime 250mg, n/'-hydroxy-2-methylpropanimidamide, propanimidamide, n-hydroxy-2-methyl
PubChem CID 9582835
IUPAC Name N'-hydroxy-2-methylpropanimidamide
SMILES CC(C)C(\N)=N/O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 102.14
Percent Purity 97%
Chemical Name or Material N';-Hydroxy-2-methylpropanimidamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.