missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy-1,8-naphthalimide, 97%, Thermo Scientific™

Product Code. 11490667
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490667 100 g 100g
11470667 5 g 5g
11480667 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490667 Supplier Thermo Scientific Alfa Aesar Supplier No. B22485.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in chemical research & chemical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7797-81-1
Molecular Formula C12H7NO3
Molecular Weight (g/mol) 213.192
MDL Number MFCD00065062
InChI Key KTWCUGUUDHJVIH-UHFFFAOYSA-N
Synonym n-hydroxynaphthalimide, n-hydroxy-1,8-naphthalimide, naphthalimide, n-hydroxy, naphthalhydroxamic acid, 2-hydroxy-1h-benzo de isoquinoline-1,3 2h-dione, n-hydroxy-1,8-naphthalenedicarboximide, 2-hydroxy-1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxybenzo de isoquinoline-1,3-dione, 1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxy, unii-dzg9avw5v2
PubChem CID 82263
IUPAC Name 2-hydroxybenzo[de]isoquinoline-1,3-dione
SMILES C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)O

Specifications

Melting Point 283°C to 286°C
Quantity 100 g
Beilstein 178595
Solubility Information Insoluble in water.
Formula Weight 213.19
Percent Purity 97%
Chemical Name or Material N-Hydroxy-1,8-naphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.