missing translation for 'onlineSavingsMsg'
Learn More

n-Hexylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15465355
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465355 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15465355 Supplier Thermo Scientific Alfa Aesar Supplier No. H58384.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 124397-96-2
Density 0.965
Molecular Formula C6H13BrZn
Linear Formula CH3(CH2)5ZnBr
MDL Number MFCD00671999
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 1-hexylzinc bromide, bromo hexyl zinc, hexylzinc bromide solution, 0.5 m in thf, 1-hexylzinc bromide 0.5 m in tetrahydrofuran, n-hexylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
InChI Key IWFHUAMVSWOCPN-UHFFFAOYSA-M
SMILES CCCCC[CH2-].[Zn+]Br
IUPAC Name bromozinc(1+);hexane
Molecular Weight (g/mol) 230.454
PubChem CID 3321884
Formula Weight 230.45
Chemical Name or Material n-Hexylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.