missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyltrimethoxysilane, 97%

Product Code. 11322226
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322226 25 g 25g
11312226 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322226 Supplier Thermo Scientific Alfa Aesar Supplier No. L03576.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3069-19-0 | C9H22O3Si | 206.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3069-19-0
Molecular Formula C9H22O3Si
Molecular Weight (g/mol) 206.36
MDL Number MFCD00039801
InChI Key CZWLNMOIEMTDJY-UHFFFAOYSA-N
Synonym n-hexyltrimethoxysilane, silane, hexyltrimethoxy, unii-9aui936hic, hexyl trimethoxy silane, 9aui936hic, hexyl-trimethoxysilane, acmc-209hhw, czwlnmoiemtdjy-uhfffaoysa
PubChem CID 76478
IUPAC Name hexyl(trimethoxy)silane
SMILES CCCCCC[Si](OC)(OC)OC

Specifications

Boiling Point 202°C to 203°C
Quantity 25 g
Beilstein 2347551
Flash Point 62°C (143°F)
Formula Weight 206.36
Percent Purity 97%
Refractive Index 1.407
Sensitivity Moisture sensitive
Density 0.92
Chemical Name or Material n-Hexyltrimethoxysilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.