missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyltriethoxysilane, 97%

Product Code. 11359215
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359215 25 g 25g
11369215 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359215 Supplier Thermo Scientific Alfa Aesar Supplier No. L16521.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18166-37-5 | C12H28O3Si | 248.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18166-37-5
Molecular Formula C12H28O3Si
Molecular Weight (g/mol) 248.44
MDL Number MFCD00087744
InChI Key WUMSTCDLAYQDNO-UHFFFAOYSA-N
Synonym hexyltriethoxysilane, triethoxy hexyl silane, hexyl triethoxysilane, n-hexyltriethoxysilane, hexyl-triethoxysilane, acmc-209ei3
PubChem CID 4437318
IUPAC Name triethoxy(hexyl)silane
SMILES CCCCCC[Si](OCC)(OCC)OCC

Specifications

Boiling Point 115°C (18mmHg)
Quantity 25 g
Linear Formula CH3(CH2)5Si(OCH2CH3)3
Beilstein 1757895
Flash Point 95°C (203°F)
Formula Weight 248.44
Percent Purity 97%
Sensitivity Moisture sensitive
Density 0.86
Chemical Name or Material n-Hexyltriethoxysilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.