missing translation for 'onlineSavingsMsg'
Learn More

n-Hexylbenzene, 98%

Product Code. 11435203
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435203 500 g 500g
11415203 25 g 25g
11425203 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435203 Supplier Thermo Scientific Alfa Aesar Supplier No. A12821.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1077-16-3 | C12H18 | 162.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1077-16-3
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00009526
InChI Key LTEQMZWBSYACLV-UHFFFAOYSA-N
Synonym 1-phenylhexane, n-hexylbenzene, benzene, hexyl, hexane, 1-phenyl, unii-8e3d1a2bzp, 8e3d1a2bzp, phenylhexane, hexyl-benzen, 1-hexylbenzene, 4-hexylbenzene
PubChem CID 14109
IUPAC Name hexylbenzene
SMILES CCCCCCC1=CC=CC=C1

Specifications

Melting Point -61°C
Density 0.861
Boiling Point 226°C to 227°C
Flash Point 83°C (181°F)
Refractive Index 1.486
Quantity 500 g
UN Number UN3082
Beilstein 1904786
Formula Weight 162.28
Percent Purity 98%
Chemical Name or Material n-Hexylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.