missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyl formate, 97%, Thermo Scientific™

Product Code. 11332426
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332426 250 g 250g
11322426 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332426 Supplier Thermo Scientific Alfa Aesar Supplier No. L06226.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-33-4
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00046146
InChI Key OUGPMNMLWKSBRI-UHFFFAOYSA-N
Synonym n-hexyl formate, formic acid, hexyl ester, n-hexyl methanoate, hexyl methanoate, formic acid hexyl ester, hexyl formiate, unii-9gpu7pgv1g, fema no. 2570, 9gpu7pgv1g, hexylformate
PubChem CID 61177
IUPAC Name hexyl formate
SMILES CCCCCCOC=O

Specifications

Melting Point -63°C
Density 0.875
Boiling Point 155°C
Flash Point 37°C (98°F)
Refractive Index 1.408
Quantity 250 g
UN Number UN3272
Beilstein 1701629
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material n-Hexyl formate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.