missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyl benzoate, 98+%

Product Code. 11312456
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312456 25 g 25g
11322456 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312456 Supplier Thermo Scientific Alfa Aesar Supplier No. L06293.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6789-88-4 | C13H18O2 | 206.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6789-88-4
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00051714
InChI Key UUGLJVMIFJNVFH-UHFFFAOYSA-N
Synonym benzoic acid, hexyl ester, benzoic acid n-hexyl ester, n-hexylbenzoate, n-hexyl benzoate, 1-hexyl benzoate, hexylbenzoate, hexylester kyseliny benzoove, benzoic acid hexyl ester, unii-vr3c36ekjr, fema no. 3691
PubChem CID 23235
IUPAC Name hexyl benzoate
SMILES CCCCCCOC(=O)C1=CC=CC=C1

Specifications

Density 0.98
Boiling Point 272°C
Flash Point 148°C (298°F)
Refractive Index 1.493
Quantity 25 g
Beilstein 2048117
Formula Weight 206.29
Percent Purity ≥98%
Chemical Name or Material n-Hexyl benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.