missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyl acetate, 99%

Product Code. 11362476
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362476 250 mL 250mL
11372476 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362476 Supplier Thermo Scientific Alfa Aesar Supplier No. L06342.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-92-7 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
n-Hexyl acetate exhibits antimicrobial activity and can be used to improve the safety of minimally processed fruits. It is a fruity smelling fluid used as flavoring agent or in perfumes. It is a green leaf volatile from cabbage Brassica oleracea var. capitata L. It is used as plasticizers.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Store at 4°C. Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-92-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00009524
InChI Key AOGQPLXWSUTHQB-UHFFFAOYSA-N
Synonym n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural
PubChem CID 8908
ChEBI CHEBI:87510
IUPAC Name hexyl acetate
SMILES CCCCCCOC(=O)C

Specifications

Melting Point -80°C
Density 0.873
Boiling Point 167°C to 169°C
Flash Point 43°C (109°F)
Linear Formula CH3CO2(CH2)5CH3
Refractive Index 1.409
Quantity 250 mL
UN Number UN3271
Beilstein 1747138
Solubility Information Soluble in alcohol,ether. Insoluble in water.
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material n-Hexyl acetate
Show More Show Less
compliance-icons
Product Identifier
  • n-Hexyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: n-Hexyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.