missing translation for 'onlineSavingsMsg'
Learn More

n-Hexatriacontane, 97+%, Thermo Scientific Chemicals

Catalog Number p-7049938
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
630-06-8
C36H74
506.99
MFCD00009413
YDLYQMBWCWFRAI-UHFFFAOYSA-N
n-hexatriacontane, hexatriacontane, ch3-ch2 34-ch3, hexatriacontane, analytical standard, hexatriacontane, n, acmc-1b3b7, n-hexatriacontane 10g
12412
CHEBI:72688
hexatriacontane
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
630-06-8
C36H74
506.99
MFCD00009413
YDLYQMBWCWFRAI-UHFFFAOYSA-N
n-hexatriacontane, hexatriacontane, ch3-ch2 34-ch3, hexatriacontane, analytical standard, hexatriacontane, n, acmc-1b3b7, n-hexatriacontane 10g
12412
CHEBI:72688
hexatriacontane
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

n-Hexatriacontane is used as a paraffin wax and in candles. It is used as a key starting material for the preparation of lower alkanes by cracking. It is also used for example in road surfacing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 76.2°C
Color White
Density 0.779 g/mL
Boiling Point 497°C
Refractive Index 1.4573
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 506.95
Percent Purity ≥97%
Physical Form Wax
Chemical Name or Material n-Hexatriacontane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'