missing translation for 'onlineSavingsMsg'
Learn More

n-Hexatriacontane, 97+%

Product Code. 11344167
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344167 50 g 50g
11354167 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11344167 Supplier Thermo Scientific Alfa Aesar Supplier No. 031641.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 630-06-8 | C36H74 | 506.99 g/mol

n-Hexatriacontane is used as a paraffin wax and in candles. It is used as a key starting material for the preparation of lower alkanes by cracking. It is also used for example in road surfacing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-06-8
Molecular Formula C36H74
Molecular Weight (g/mol) 506.99
MDL Number MFCD00009413
InChI Key YDLYQMBWCWFRAI-UHFFFAOYSA-N
Synonym n-hexatriacontane, hexatriacontane, ch3-ch2 34-ch3, hexatriacontane, analytical standard, hexatriacontane, n, acmc-1b3b7, n-hexatriacontane 10g
PubChem CID 12412
ChEBI CHEBI:72688
IUPAC Name hexatriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 76.2°C
Color White
Density 0.779 g/mL
Boiling Point 497°C
Refractive Index 1.4573
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 506.95
Percent Purity ≥97%
Physical Form Wax
Chemical Name or Material n-Hexatriacontane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.