Product Code. 16665893
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
16665893
2.5 L
2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
16665893
Supplier
Chromasolv
Supplier No.
657302.5L
250.80 GBP valid until 2026-10-31 BEST PRICE on Promo! Use promo code "27992" to get your promotional price.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Specifications
Melting Point
-95°C
CAS
110-54-3
Color
Colorless
CAS Max %
1
Density
0.660 - 0.668 g/cm3 (20°C)
Quantity
2.5 L
Flash Point
-26°C
Molecular Formula
C6H14
Linear Formula
CH3 (CH2 )4 CH3
MDL Number
MFCD02179311
Boiling Point
66°C to 69°C (1013 hPa)
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES
CCCCCC
IUPAC Name
hexane
Molecular Weight (g/mol)
86.18
Chemical Name or Material
n-Hexane
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?