missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, 99%, ChromAR™ Solvent, Macron Fine Chemicals™

Product Code. 15528724
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15528724 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15528724 Supplier Macron Fine Chemicals Supplier No. 671325
  • ONLINE EXCLUSIVE PRICE
    £530.00
    £397.24 / 2.5 L
    Save £132.76
    25% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

High-purity solvent that meets ACS specifications and is suitable for liquid chromatography and UV-spectrophotometry applications.

Macron Fine Chemicals brand ChromAR™ grade solvents are ideal for use in analytical testing in a wide variety of markets where consistency and reliability is required. These reagents are produced under strict manufacturing practices for a variety of liquid chromatography applications.

  • Consistent gradient profiles for reproducible results—ideal for consistent mobile phases
  • Low UV absorbance
  • Flat baselines
  • Low residue after evaporation
  • Low water levels

Specifications

Melting Point -95°C
Absorbance Max. 0.15 (at 210nm), Max. 0.05 (at 220nm), Max. 0.02 (at 230nm), Max. 0.009 (at 240nm)
CAS 110-54-3
Color Colorless
Density 0.66g/cm3 (at 20°C)
Quantity 2.5 L
Flash Point −22°C
Ignition Point 240°C
Physical Form Liquid
Residue after Evaporation ≤0.0005%
Titratable Acid ≤0.0005meq/g
Molecular Formula C6H14
Titratable Base ≤0.0003meq/g
MDL Number MFCD02179311
Viscosity 0.326 mPas (dynamic), 0.50mm2/s (kinematic)
Water ≤0.01%
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
Boiling Point 69°C
Solubility Information 0.1g/L in water
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES CCCCCC
IUPAC Name hexane
Molecular Weight (g/mol) 86.18
PubChem CID 8058
ChEBI CHEBI:29021
Percent Purity ≥99%
Grade Suitable for HPLC, UV-Spectrophotometry and UV applications
Chemical Name or Material n-Hexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.