missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecyl mercaptan, 90%, technical, ACROS Organics™

Brand:  Acros Organics 120520100

Code : 29

Additional Details : CAS Number : 2917-26-2 Weight : 0.05840kg

Packaging Glass bottle
Quantity 10mL

 View more versions of this product

Product Code. 10041090

  • £15.58 / 10mL

Estimated Shipment
12-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

2917-26-2
258.5
ORTRWBYBJVGVQC-UHFFFAOYSA-N
18015
CCCCCCCCCCCCCCCCS
C16H34S
MFCD00011677
1-hexadecanethiol, hexadecanethiol, n-hexadecanethiol, cetyl mercaptan, n-hexadecyl mercaptan, hexadecyl mercaptan, unii-qr98qio1ql, 1-cetanethiol, qr98qio1ql, cetylmercaptan
hexadecane-1-thiol

Specifications

n-Hexadecyl mercaptan
101°C
Glass bottle
0.84
Colorless or White to Yellow
10mL
88% min. (GC)
CH3(CH2)15SH
01, 430
Solubility in water: insoluble in water. Other solubilities: slightly soluble in ethyl alcohol,carbon tetrachl,soluble in ether
CCCCCCCCCCCCCCCCS
258.5
258.5
90%
0.8400g/mL
Authentic
1.4610 to 1.463
184°C to 191°C (7.0mmHg)
18°C to 20°C
2917-26-2
C16H34S
MFCD00011677
1-hexadecanethiol, hexadecanethiol, n-hexadecanethiol, cetyl mercaptan, n-hexadecyl mercaptan, hexadecyl mercaptan, unii-qr98qio1ql, 1-cetanethiol, qr98qio1ql, cetylmercaptan
ORTRWBYBJVGVQC-UHFFFAOYSA-N
hexadecane-1-thiol
18015
Liquid or Waxy Solid
Technical
Videos
SDS Documents