missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecyl beta-D-maltopyranoside, 98%, Thermo Scientific™

Product Code. 15497929
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497929 100 mg 100mg
15407939 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497929 Supplier Thermo Scientific Alfa Aesar Supplier No. J66531.MC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98064-96-1
Molecular Formula C28H54O11
Molecular Weight (g/mol) 566.729
MDL Number MFCD01310880
InChI Key GTQCHJYVKDXMRU-YMEALESQSA-N
Synonym n-hexadecyl, a-d-maltoside, n-hexadecyl beta-d-maltoside, hexadecyl b-d-maltopyranoside, hexadecyl beta-d-maltopyranoside, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-hexadecyloxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, cetyl, cetyl beta-d-maltoside, hexadecyl beta-maltoside, hexadecyl-, b-d-glucopyranoside, hexadecyl4-o-a-d-glucopyranosyl
PubChem CID 9850929
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-hexadecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O

Specifications

Color White
Quantity 100 mg
Sensitivity Moisture sensitive
Formula Weight 566.72
Percent Purity 98%
Physical Form Powder
Chemical Name or Material n-Hexadecyl beta-D-maltopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.