missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecane, 99%, pure

Product Code. p-4229234 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
10 L
Unit Size:
10 L
100mL
2.5 L
500mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10449610

Brand: Thermo Scientific Acros 120461000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

n-Hexadecane, 99%, C16H34, CAS Number-544-76-3, n-cetane, n-hexadecane, hexadecan, hexadekan, cetan, cetane, ccris 5833, zetan, pentadecane, methyl, unii-f8z00shp6q, 2.5L, 98.5% min. (GC), 01,172, 287.0 deg.C, 100.0, CHEBI:45296, Colorless, 0.7730g/mL, 208-878-9, 135 deg.C, 226.44, Pure | CAS: 544-76-3 | C16H34 | 226.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16H34
Molecular Weight (g/mol) 226.44
MDL Number MFCD00008998
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
Synonym n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
PubChem CID 11006
ChEBI CHEBI:45296
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 18.0°C
Color Colorless
Density 0.7730g/mL
Boiling Point 287.0°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)14CH3
Refractive Index 1.4330 to 1.4350
Quantity 100 mL
Beilstein 01,172
Specific Gravity 0.773
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone and ether
Viscosity 3.032 mPa.s (25°C)
Formula Weight 226.44
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Hexadecane
Show More Show Less
compliance-icons
Product Identifier
  • n-Hexadecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.