missing translation for 'onlineSavingsMsg'
Learn More

n-Hexacosane, 99%

Product Code. 11398968
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11398968

Brand: Thermo Scientific Alfa Aesar 042461.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 630-01-3 | C26H54 | 366.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-01-3
Molecular Formula C26H54
Molecular Weight (g/mol) 366.72
MDL Number MFCD00009354
InChI Key HMSWAIKSFDFLKN-UHFFFAOYSA-N
Synonym n-hexacosane, pentacosane, methyl, ch3-ch2 24-ch3, hexacosane, analytical standard, hexacosane, acmc-1b86y
PubChem CID 12407
ChEBI CHEBI:32940
IUPAC Name hexacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 56°C to 58°C
Color White
Odor Weak
Quantity 5 g
Beilstein 1705609
Solubility Information Slightly soluble in water.
Formula Weight 366.72
Percent Purity 99%
Physical Form Solid
Chemical Name or Material n-Hexacosane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.