missing translation for 'onlineSavingsMsg'
Learn More

n-Heptyl 4-Hydroxybenzoate, TRC

Product Code. 30393082
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393082 5 g 5g
30406320 100 g -
30389922 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393082 Supplier Toronto Research Chemicals Supplier No. TRCH2831805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1085-12-7
Molecular Formula C14 H20 O3
Molecular Weight (g/mol) 236.31
Synonym Benzoic acid, 4-hydroxy-, heptyl ester (9CI, ACI), Benzoic acid, p-hydroxy-, heptyl ester (6CI, 7CI, 8CI), Heptyl alcohol, p-hydroxybenzoate (6CI), Heptyl 4-hydroxybenzoate, Heptyl p-hydroxybenzoate, Heptyl paraben, n-Heptyl 4-hydroxybenzoate, n-Heptyl p-hydroxybenzoate, Nipaheptyl, NSC 309818, Staypro WS 7, 4-Hydroxybenzoic acid heptyl ester
IUPAC Name heptyl 4-hydroxybenzoate
SMILES CCCCCCCOC(=O)c1ccc(O)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H20O3/c1-2-3-4-5-6-11-17-14(16)12-7-9-13(15)10-8-12/h7-10,15H,2-6,11H2,1H3
Formula Weight 236.1412
Percent Purity >95%
Chemical Name or Material 4-Hydroxybenzoic acid-n-heptyl ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.