missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Heptanoic Acid Tianeptine Dimethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | N-Heptanoic Acid Tianeptine Dimethyl Ester |
| Analyte or Component Names | N-Heptanoic Acid Tianeptine Dimethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C30 H41 Cl N2 O6 S |
| Synonym | Dimethyl 7,7'-((3-Chloro-6-methyl-5,5-dioxido-6,11-dihydrodibenzo[c,f][1,2]thiazepin-11-yl)azanediyl)diheptanoate |
| InChI Formula | InChI=1S/C30H41ClN2O6S/c1-32-26-15-11-10-14-24(26)30(25-19-18-23(31)22-27(25)40(32,36)37)33(20-12-6-4-8-16-28(34)38-2)21-13-7-5-9-17-29(35)39-3/h10-11,14-15,18-19,22,30H,4-9,12-13,16-17,20-21H2,1-3H3 |
| Solution Type | Neat |
| SMILES | COC(=O)CCCCCCN(CCCCCCC(=O)OC)C1c2ccccc2N(C)S(=O)(=O)c3cc(Cl)ccc13 |
| IUPAC Name | methyl 7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][1,2]benzothiazepin-11-yl)-(7-methoxy-7-oxoheptyl)amino]heptanoate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?