missing translation for 'onlineSavingsMsg'
Learn More

N-Heptanoic Acid Tianeptine Dimethyl Ester, TRC

Product Code. 30396835
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396835 5 mg 5mg
30399595 25 mg 25mg
30403711 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396835 Supplier Toronto Research Chemicals Supplier No. TRCH2815105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Heptanoic Acid Tianeptine Dimethyl Ester
Analyte or Component Names N-Heptanoic Acid Tianeptine Dimethyl Ester
Shipping Condition Room Temperature
Molecular Formula C30 H41 Cl N2 O6 S
Synonym Dimethyl 7,7'-((3-Chloro-6-methyl-5,5-dioxido-6,11-dihydrodibenzo[c,f][1,2]thiazepin-11-yl)azanediyl)diheptanoate
InChI Formula InChI=1S/C30H41ClN2O6S/c1-32-26-15-11-10-14-24(26)30(25-19-18-23(31)22-27(25)40(32,36)37)33(20-12-6-4-8-16-28(34)38-2)21-13-7-5-9-17-29(35)39-3/h10-11,14-15,18-19,22,30H,4-9,12-13,16-17,20-21H2,1-3H3
Solution Type Neat
SMILES COC(=O)CCCCCCN(CCCCCCC(=O)OC)C1c2ccccc2N(C)S(=O)(=O)c3cc(Cl)ccc13
IUPAC Name methyl 7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][1,2]benzothiazepin-11-yl)-(7-methoxy-7-oxoheptyl)amino]heptanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 593.174
Quantity 5 mg
Formula Weight 592.237
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.