missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, for GC,Solstice

Product Code. 16635913
Change view
Click to view available options
Quantity:
7 L
Unit Size:
7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16635913 7 L 7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 16635913 Supplier Chromasolv Supplier No. 658367LRC
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Specifications

Melting Point -90.5°C
CAS 142-82-5
Color Colorless
CAS Max % 1
Density 0.69 g/cm3 (15°C)
Quantity 7 L
Flash Point -4°C
Molecular Formula C7H16
Linear Formula CH3(CH2)5CH3
MDL Number MFCD00009544
Boiling Point 98.2°C to 98.4°C (1000 hPa)
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.21
Grade GC
Chemical Name or Material n-Heptane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.