Product Code. 12695047
Shop All Thermo Scientific Chemicals Products
Click to view available options
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
12695047
1 L
1 L
12662906
2.5 L
2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
12695047
Supplier
Thermo Scientific Acros
Supplier No.
447070010
£46.25 £42.70 / 1 L Save £3.55 7% Off
Estimated Shipment: 17-02-2027Log in to see stock.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
CAS: 142-82-5 | C7H16 | 100.21 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
CAS
142-82-5
Molecular Formula
C7H16
Molecular Weight (g/mol)
100.21
MDL Number
MFCD00009544
InChI Key
IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID
8900
ChEBI
CHEBI:43098
IUPAC Name
heptane
SMILES
CCCCCCC
Spécification
Melting Point
-91.0°C
Density
0.6830g/mL
Boiling Point
98.0°C
Flash Point
−4°C
Assay Percent Range
98.5% min. (GC)
Linear Formula
CH3 (CH2 )5 CH3
Refractive Index
1.3850 to 1.3890
Quantity
1 L
Merck Index
15,4694
Specific Gravity
0.683
Solubility Information
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Viscosity
0.4 mPa.s (20°C)
Formula Weight
100.2
Percent Purity
99%
Grade
Biochemistry
Chemical Name or Material
n-Heptane
Afficher plus
Afficher moins
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité.
Politique de confidentialité .
Vous avez repéré une opportunité d'amélioration ?