missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane ≥ 99.5%, ChromAR™ for HPLC, Fisher Chemical™

Product Code. 15733872
Change view
Click to view available options
:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15733872 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15733872 Supplier Macron Fine Chemicals Supplier No. 513925

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Suitable for Liquid Chromatography and UV-Spectrophotometry

  • Ultraviolet Absorbance (1.00-cm cell vs water):
    • at 210 nm max. 1.0
    • at 220 nm max. 0.1
    • at 230 nm max. 0.02
    • at 240 nm max. 0.01
    • at 250 nm max. 0.005

Specifications

CAS 142-82-5
Acidity max. 0.0005 meq/g
Density 0.68 g/mL
Quantity 2.5 L
Flash Point -4°C
Packaging Glass Bottle
Residue after Evaporation max. 0.0005%
Assay Percent Range min. 99.5%
Molecular Formula C7H16
MDL Number MFCD00009544
UN Number 1206
Water max. 0.01%
Boiling Point 98°C
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.21
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.