missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, 99.5%, for analysis

Product Code. 10658614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Unit Size:
1 L
2.5 L
25 L
5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10658614 5 L 5 L
10215463 1 L 1 L
10139442 2.5 L 2.5 L
13157754 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10658614 Supplier Thermo Scientific Acros Supplier No. 389170050
  • ONLINE EXCLUSIVE PRICE
    £228.00
    £221.00 / 5 L
    Save £7.00
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 142-82-5 | C7H16 | 100.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -91.0°C
CAS Min % 99.4
Acidity 0.0001 meq/g max.
CAS Max % 100.0
Density 0.6830g/mL
Quantity 5 L
Flash Point −4°C
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.3850 to 1.3890
Residue after Evaporation 0.001% max.
Assay Percent Range 99.5%
Specific Gravity 0.683
Linear Formula CH3(CH2)5CH3
Viscosity 0.4 mPa.s (20°C)
Water 0.02% max. (K.F.)
Boiling Point 98.0°C
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 100.2
Percent Purity 99.5%
Grade Analytical
Chemical Name or Material n-Heptane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Heptane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.