missing translation for 'onlineSavingsMsg'
Learn More

N-Heptafluorobutyryl-Imidazole, 98+%, Pure, Fisher Chemical

Catalog Number 10794941
Click to view available options
Quantity:
5 g
Packaging:
Plastic container
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 32477-35-3
Melting Point 10°C
Color Colorless
Boiling Point 158°C
Packaging Plastic container
Molecular Formula C7H3F7N2O
MDL Number 14503
Quantity 5 g
Synonym n-heptafluorobutyrylimidazole, 1-heptafluorobutyryl imidazole, heptafluorobutyrylimidazole, n-heptafluorobutyryl imidazole, hfbi, 1-perfluorobutyryl imidazole, 2,2,3,3,4,4,4-heptafluoro-1-1h-imidazol-1-yl butan-1-one, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl-1h-imidazole, 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-yl butan-1-one, 1h-imidazole, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl
InChI Key MSYHGYDAVLDKCE-UHFFFAOYSA-N
SMILES C1=CN(C=N1)C(=O)C(C(C(F)(F)F)(F)F)(F)F
IUPAC Name 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-ylbutan-1-one
Molecular Weight (g/mol) 264.103
PubChem CID 94431
Formula Weight 264.1g/mol
Physical Form Liquid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'