missing translation for 'onlineSavingsMsg'
Learn More

n-Heptadecane, 99%

Product Code. 11343907
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343907 25 g 25g
11333907 100 g 100g
11323907 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11343907 Supplier Thermo Scientific Alfa Aesar Supplier No. 031551.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 629-78-7 | C17H36 | 240.475 g/mol

n-Heptadecane is used as an internal standard to study the first direct comparison of kava lactone extraction efficiencies. It plays an important role as a suspended solvent used for the extraction and concentration of essential oil. It is used as a precursor in the production of iminoctadine triacetate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-78-7
Molecular Formula C17H36
Molecular Weight (g/mol) 240.475
MDL Number MFCD00009002
InChI Key NDJKXXJCMXVBJW-UHFFFAOYSA-N
Synonym n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078
PubChem CID 12398
ChEBI CHEBI:16148
IUPAC Name heptadecane
SMILES CCCCCCCCCCCCCCCCC

Specifications

Melting Point 22°C
Density 0.778
Boiling Point 302°C
Refractive Index 1.4369
Quantity 25 g
Solubility Information Immiscible with water.
Formula Weight 240.48
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material n-Heptadecane
compliance-icons
Produktidentifikator
  • n-Heptadecane
Signalord
  • Danger
Farekategori
  • Aspiration toxicity Category 1
Faresætning
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplerende oplysninger
  • MIXTURE LIST-Contain: Heptadecane

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.