missing translation for 'onlineSavingsMsg'
Learn More

n-Heptadecane, 99.5%

Product Code. 11425250
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425250 5 g 5g
11415250 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425250 Supplier Thermo Scientific Alfa Aesar Supplier No. 044514.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 629-78-7 | C17H36 | 240.475 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-78-7
Molecular Formula C17H36
Molecular Weight (g/mol) 240.475
MDL Number MFCD00009002
InChI Key NDJKXXJCMXVBJW-UHFFFAOYSA-N
Synonym n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078
PubChem CID 12398
ChEBI CHEBI:16148
IUPAC Name heptadecane
SMILES CCCCCCCCCCCCCCCCC

Specifications

Melting Point 22°C
Density 0.7783 g/mL
Boiling Point 302°C
Refractive Index 1.4369
Quantity 5 g
Formula Weight 240.48
Percent Purity 99.5%
Physical Form Crystalline
Chemical Name or Material n-Heptadecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.