missing translation for 'onlineSavingsMsg'
Learn More

n-Heneicosane, 98%

Product Code. 10086053 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10086053 50 g Glass bottle 50g
10602952 1 g Glass bottle 1g
10182951 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10086053 Supplier Thermo Scientific Acros Supplier No. 352770500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 629-94-7 | C21H44 | 296.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-94-7
Molecular Formula C21H44
Molecular Weight (g/mol) 296.58
MDL Number MFCD00009346
InChI Key FNAZRRHPUDJQCJ-UHFFFAOYSA-N
Synonym heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj
PubChem CID 12403
ChEBI CHEBI:32931
IUPAC Name henicosane
SMILES CCCCCCCCCCCCCCCCCCCCC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 39°C to 43°C
Color White
Boiling Point 100°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)19CH3
Quantity 50 g
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,,soluble in petroleum ether
Formula Weight 296.58
Percent Purity 98%
Physical Form Crystalline Chunks, Crystals, or Flakes
Chemical Name or Material n-Heneicosane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.