missing translation for 'onlineSavingsMsg'
Learn More

n-Heneicosane, 98%

Product Code. p-3778739 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10602952

Brand: Thermo Scientific 352770010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 629-94-7 | C21H44 | 296.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-94-7
Molecular Formula C21H44
Molecular Weight (g/mol) 296.58
MDL Number MFCD00009346
InChI Key FNAZRRHPUDJQCJ-UHFFFAOYSA-N
Synonym heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj
PubChem CID 12403
ChEBI CHEBI:32931
IUPAC Name henicosane
SMILES CCCCCCCCCCCCCCCCCCCCC

Specifica

CAS Min % 97.5
CAS Max % 100.0
Melting Point 39°C to 43°C
Color White
Boiling Point 100°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)19CH3
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,,soluble in petroleum ether
Formula Weight 296.58
Percent Purity 98%
Physical Form Crystalline Chunks, Crystals, or Flakes
Chemical Name or Material n-Heneicosane
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato