missing translation for 'onlineSavingsMsg'
Learn More

N-Guanylurea sulfate, 97%

Product Code. 11449406
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449406 250 g 250g
11429406 10 g 10g
11439406 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449406 Supplier Thermo Scientific Alfa Aesar Supplier No. A19106.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-01-5 | C4H12N8O2·H2SO4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-01-5
Molecular Formula C4H12N8O2·H2SO4
MDL Number MFCD00035565
Synonym Dicyanodiamidine sulfate

Specifications

Melting Point ∼195°C (decomposition)
Quantity 250 g
Beilstein 6099371
Merck Index 14,3093
Formula Weight 302.27
Percent Purity 97%
Chemical Name or Material N-Guanylurea sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.