missing translation for 'onlineSavingsMsg'
Learn More

N-Glycyl-L-tyrosine, 98%

Product Code. p-7056460 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 17113076

Brand: Thermo Scientific H6538822

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 658-79-7 | C11H14N2O4 | 238.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 658-79-7
Molecular Formula C11H14N2O4
Molecular Weight (g/mol) 238.24
MDL Number MFCD00008126
InChI Key XBGGUPMXALFZOT-IMWMWJONNA-N
Synonym glycyl-l-tyrosine, gly-tyr, n-glycyl-l-tyrosine, h-gly-tyr-oh, glycyltyrosine, l-tyrosine, glycyl, s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid, chembl53400, 2s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2-aminoacetyl amino-3-4-hydroxyphenyl propanoic acid
PubChem CID 92829
ChEBI CHEBI:73517
IUPAC Name (2S)-2-(2-aminoacetamido)-3-(4-hydroxyphenyl)propanoic acid
SMILES NCC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White to Yellow
Beilstein 2700715
Quantity 100 g
Formula Weight 238.24
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Glycyl-L-tyrosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.