missing translation for 'onlineSavingsMsg'
Learn More

N-Glycyl-L-tyrosine, 98%

Product Code. 17113076 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17113076 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17113076 Supplier Thermo Scientific Supplier No. H6538822
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 658-79-7 | C11H14N2O4 | 238.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 658-79-7
Molecular Formula C11H14N2O4
Molecular Weight (g/mol) 238.24
MDL Number MFCD00008126
InChI Key XBGGUPMXALFZOT-IMWMWJONNA-N
Synonym glycyl-l-tyrosine, gly-tyr, n-glycyl-l-tyrosine, h-gly-tyr-oh, glycyltyrosine, l-tyrosine, glycyl, s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid, chembl53400, 2s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2-aminoacetyl amino-3-4-hydroxyphenyl propanoic acid
PubChem CID 92829
ChEBI CHEBI:73517
IUPAC Name (2S)-2-(2-aminoacetamido)-3-(4-hydroxyphenyl)propanoic acid
SMILES NCC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Especificaciones

Color White to Yellow
Beilstein 2700715
Quantity 100 g
Formula Weight 238.24
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Glycyl-L-tyrosine

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.