missing translation for 'onlineSavingsMsg'
Learn More

N-Formylnornicotine, TRC

Product Code. 30390428
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390428 25 mg
30406792 50 mg
30397228 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390428 Supplier Toronto Research Chemicals Supplier No. TRCF70087525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3000-81-5
Molecular Formula C11 H14 N2 O
Molecular Weight (g/mol) 190.24
IUPAC Name 2-pyridin-3-ylpiperidine-1-carbaldehyde
SMILES O=CN1CCCCC1c2cccnc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H14N2O/c14-9-13-7-2-1-5-11(13)10-4-3-6-12-8-10/h3-4,6,8-9,11H,1-2,5,7H2
Formula Weight 190.1106
Chemical Name or Material N-Formylnornicotine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.