missing translation for 'onlineSavingsMsg'
Learn More

N-Formylmorpholine, 99%

Product Code. p-137008 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
This item is not returnable. View return policy

Product Code. 10346540

Brand: Thermo Scientific Acros 133800050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4394-85-8 | C5H9NO2 | 115.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4394-85-8
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00006170
InChI Key LCEDQNDDFOCWGG-UHFFFAOYSA-N
Synonym 4-formylmorpholine, n-formylmorpholine, 4-morpholinecarboxaldehyde, morpholine, 4-formyl, n-formylmorfolin, 4-morpholinecarbaldehyde, n-formylmorfolin czech, unii-d1e63xo4rh, n-formyhnorpholine, n-formyl morpholin
PubChem CID 20417
ChEBI CHEBI:43989
IUPAC Name morpholine-4-carbaldehyde
SMILES C1COCCN1C=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 23°C
Color Yellow
Density 1.1400g/mL
Boiling Point 240°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4855 to 1.4875
Quantity 5 mL
Specific Gravity 1.14
Solubility Information Solubility in water: soluble in water
Viscosity 6.79 mPa.s (30°C)
Formula Weight 115.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N-Formylmorpholine, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.