missing translation for 'onlineSavingsMsg'
Learn More

N-Formylguanidine, TRC

Product Code. 30397640
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397640 1 g 1g
30391748 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397640 Supplier Toronto Research Chemicals Supplier No. TRCF7005051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4471-51-6
Molecular Formula C2H5N3O
Molecular Weight (g/mol) 87.08
Synonym N-Amidinoformamide, 1-Formyl-3-cyanoguanidine, N-(Aminoiminomethyl)formamide, Formylguanidine, NSC 77929
IUPAC Name N-carbamimidoylformamide
SMILES NC(NC=O)=N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H5N3O/c3-2(4)5-1-6/h1H,(H4,3,4,5,6)
Formula Weight 87.04
Chemical Name or Material N-Formylguanidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.