missing translation for 'onlineSavingsMsg'
Learn More

N-Formylglycine, 98%

Product Code. 15474027
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15474027

Brand: Thermo Scientific Alfa Aesar H31053.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2491-15-8 | C3H5NO3 | 103.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-15-8
Molecular Formula C3H5NO3
Molecular Weight (g/mol) 103.08
MDL Number MFCD00037360
InChI Key UGJBHEZMOKVTIM-UHFFFAOYSA-N
Synonym n-formylglycine, formylglycine, glycine, n-formyl, fgly, for-gly-oh, formylglycin, n-hydroxymethyleneglycine, formylamino acetic acid, n-formylglycin, n-formyl-glycine
PubChem CID 75606
IUPAC Name 2-formamidoacetic acid
SMILES OC(=O)CNC=O

Specifications

Melting Point 147°C to 151°C
Linear Formula HCONHCH2CO2H
Quantity 1 g
Formula Weight 103.08
Percent Purity 98%
Chemical Name or Material N-Formylglycine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.