missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl Varenicline, TRC

Product Code. 30399837
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399837 25 mg
30408672 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399837 Supplier Toronto Research Chemicals Supplier No. TRCF70165025MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 796865-82-2
Molecular Formula C14 H13 N3 O
Molecular Weight (g/mol) 239.27
Synonym 6,10-Methano-8H-pyrazino[2,3-h][3]benzazepine-8-carboxaldehyde, 6,7,9,10-tetrahydro-, 6,7,9,10-Tetrahydro-6,10-methano-8H-pyrazino[2,3-h][3]benzazepine-8-carboxaldehyde, N-Formylvarenicline
SMILES O=CN1CC2CC(C1)c3cc4nccnc4cc23

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H13N3O/c18-8-17-6-9-3-10(7-17)12-5-14-13(4-11(9)12)15-1-2-16-14/h1-2,4-5,8-10H,3,6-7H2
Formula Weight 239.1059
Chemical Name or Material N-Formyl Varenicline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.