missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl Rifuzole, TRC

Product Code. 30396566
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396566 10 mg 10mg
30395565 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396566 Supplier Toronto Research Chemicals Supplier No. TRCF70095810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Formyl Rifuzole
Analyte or Component Names N-Formyl Rifuzole
Shipping Condition Room Temperature
Molecular Formula C9 H5 F3 N2 O2 S
Synonym N-(6-(Trifluoromethoxy)benzo[d]thiazol-2-yl)formamide
InChI Formula InChI=1S/C9H5F3N2O2S/c10-9(11,12)16-5-1-2-6-7(3-5)17-8(14-6)13-4-15/h1-4H,(H,13,14,15)
Solution Type Neat
SMILES FC(F)(F)Oc1ccc2nc(NC=O)sc2c1
IUPAC Name N-(6-(Trifluoromethoxy)benzo[d]thiazol-2-yl)formamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 262.208
Quantity 10 mg
Formula Weight 262.0024
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.